Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00948

Sigma-Aldrich

Ethylenediammonium dichloride

for synthesis

Synonym(s):

Ethylenediammonium dichloride, 1,2-Diaminoethane dihydrochloride, Ethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H8N2 · 2HCl
CAS Number:
Molecular Weight:
133.02
MDL number:
UNSPSC Code:
12352107
EC Index Number:
206-369-6
NACRES:
NA.22

Quality Level

form

powder

potency

3250 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

5 (20 °C, 100 g/L in H2O)

mp

>300 °C

solubility

300 g/L

bulk density

650 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[Cl-].NCCN.[H+].[H+]

InChI

1S/C2H8N2.2ClH/c3-1-2-4;;/h1-4H2;2*1H

InChI key

OHHBFEVZJLBKEH-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service