Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.20879

Sigma-Aldrich

2′-Nitroacetanilide

for synthesis

Synonym(s):

2′-Nitroacetanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2O3
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352111
EC Index Number:
209-009-6
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

mp

90-93 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1c(cccc1)NC(=O)C

InChI

1S/C8H8N2O3/c1-6(11)9-7-4-2-3-5-8(7)10(12)13/h2-5H,1H3,(H,9,11)

InChI key

BUNFNRVLMKHKIT-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 90 °C
Melting range (upper value): ≤ 93 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service