Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695041

Sigma-Aldrich

Nitric acid

ACS reagent, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
38060304
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

1.3 (vs air)

vapor pressure

6.6 mmHg ( 20 °C)
8 mmHg ( 20 °C)

description

dilution test passes test

Assay

≥90.0%

form

liquid

ign. residue

≤0.002%

color

colorless to yellow

bp

120.5 °C (lit.)

density

1.48 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤0.7 ppm
dissolved oxides: ≤0.1% (as N2O3)
sulfate (SO42-): ≤5 ppm

cation traces

As: ≤0.3 ppm
Fe: ≤2 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitric acid is generally used as a primary reagent for the nitration and oxidation of organic molecules. It is also used as a strong oxidizing agent in organic synthesis to perform oxidative transformations. In addition, nitric acid is also used in the manufacturing of intermediate for polyurethanes and polyamides production.

Application

Nitric acid is used:
  • As an oxidizing agent with sulfuric acid to graft the carboxylic acid over CNT (carbon nanotubes) to produce CNT-COOH.
  • In the combustion method to adjust the pH of the solution to 7. It is one of the steps in the preparation of mixed magnetite–hematite nanoparticles.
  • As a nitrating agent with sulfuric acid in the synthesis of 8-nitro-7-hydroxy-4-methyl coumarin and 6-nitro-7-hydroxy-4-methyl coumarin from 7-hydroxy-4-methyl coumarin.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren E Kremer et al.
Journal of inorganic biochemistry, 147, 227-234 (2015-05-11)
Uncontrolled reactions in biological media are a main obstacle for clinical translation of V-based anti-diabetic or anti-cancer pro-drugs. We investigated the use of controlled-release pharmaceutical formulations to ameliorate this issue with a series of V(V) and (IV) complexes of anionic
Alyson M Baergen et al.
Environmental science & technology, 47(2), 815-820 (2012-12-15)
The fate of NO(x) (=NO + NO(2)) is important to understand because NO(x) is a significant player in air quality determination through its role in O(3) formation. Here we show that renoxification of the urban atmosphere may occur through the
Stijn Smulders et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(1), 132-140 (2014-06-14)
The unique physical and chemical properties of nanomaterials have led to their increased use in many industrial applications, including as a paint additive. For example, titanium dioxide (TiO2) engineered nanoparticles (ENPs) have well-established anti-UV, self-cleaning, and air purification effects. Silver
Paul A Adlard et al.
Neurobiology of disease, 81, 196-202 (2015-01-01)
Zinc transporter-3 (ZnT3) protein is responsible for loading zinc into presynaptic vesicles and consequently controls the availability of zinc at the glutamatergic synapse. ZnT3 has been shown to decline with age and in Alzheimer's disease (AD) and is crucially involved
V Melis et al.
Cellular and molecular life sciences : CMLS, 72(11), 2199-2222 (2014-12-20)
A poorly understood feature of the tauopathies is their very different clinical presentations. The frontotemporal lobar degeneration (FTLD) spectrum is dominated by motor and emotional/psychiatric abnormalities, whereas cognitive and memory deficits are prominent in the early stages of Alzheimer's disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service