Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20832

Sigma-Aldrich

1-Methylpyrrole

for synthesis

Synonym(s):

1-Methylpyrrole, N-Methylpyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N
CAS Number:
Molecular Weight:
81.12
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-513-7
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

400 °C

potency

1400 mg/kg LD50, oral (Rat)
>400 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-8.5 % (v/v)

pH

10.4 (10 °C, 10 g/L in H2O)

bp

111-113 °C/1013 hPa

mp

-57 °C

transition temp

flash point 16 °C

solubility

13 g/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[n]1(cccc1)C

InChI

1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3

InChI key

OXHNLMTVIGZXSG-UHFFFAOYSA-N

Application

  • Systematic Assessment of the Catalytic Reactivity of Frustrated Lewis Pairs in CH Bond Activation: The article explores the FLP-catalyzed CH bond activation of 1-methylpyrrole, evaluating its catalytic performance (Y Guo, et al., 2023).
  • Design of a Mesoionic Janus-type Dicarbene: Research on 1-methylpyrrole with 4-bromoacetophenone for potential applications in synthetic chemistry (JN Leung, HV Huynh, 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.909 - 0.910
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service