Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49817

Supelco

Pyrrole

analytical standard

Synonym(s):

Azole, Divinylenimine, Imidole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N
CAS Number:
Molecular Weight:
67.09
Beilstein:
1159
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.31 (vs air)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤98.0% water

refractive index

n20/D 1.508 (lit.)
n20/D 1.508-1.511

bp

131 °C (lit.)

mp

−23 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

c1cc[nH]c1

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyrrole is used as one of the building blocks of biochemical macrocycles such as chlorins, porphyrins and corrins. It is also used for many technologically relevel applications such as in organometallic complexes and precursor molecules for self-assembled monolayers on metal surfaces.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studying Complex Surface Dynamical Systems Using Helium-3 Spin-Echo Spectroscopy (2014)
Jaehyun Hur et al.
ACS nano, 8(10), 10066-10076 (2014-09-27)
Conductive hydrogels are a class of composite materials that consist of hydrated and conducting polymers. Due to the mechanical similarity to biointerfaces such as human skin, conductive hydrogels have been primarily utilized as bioelectrodes, specifically neuroprosthetic electrodes, in an attempt
Jing Fan et al.
Journal of chromatography. A, 1362, 231-240 (2014-09-01)
Calix[4]pyrroles offer a great potential as stationary phases for gas chromatography (GC) due to their unique structures and physicochemical properties. Herein we present the first report of using two calix[4]pyrroles, namely meso-tetra-cyclohexylcalix[4]pyrrole (THCP) and meso-octamethylcalix[4]pyrrole (OMCP). These stationary phases were
SeungHyun Jeon et al.
Theranostics, 4(11), 1123-1132 (2014-09-25)
We have developed a conductive nano-roughened microfluidic device and demonstrated its use as an electrically modulated capture and release system for studying rare circulating tumor cells (CTCs). The microchannel surfaces were covalently decorated with epithelial cancer-specific anti-EpCAM antibody by electrochemical
Woo Young Hong et al.
Biomaterials, 35(36), 9573-9580 (2014-09-07)
Here, we propose an integrated multifunctional system constructed by conductive disulfide-biotin-doped polypyrrole nanowires (SS-biotin-Ppy NWs) for capture, release, and in situ quantification of circulating tumor cells (CTCs). A well-ordered three-dimensional nanowire structure equipped with a monoclonal antibody offers a significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service