Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.19016

Sigma-Aldrich

DL-Mandelic acid nitrile

for synthesis

Synonym(s):

DL-Mandelic acid nitrile, Benzaldehyde cyanohydrin, DL-α-Hydroxyphenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
12352117
EC Index Number:
208-532-7
NACRES:
NA.22

Quality Level

form

liquid

potency

116 mg/kg LD50, oral (Rat)

bp

170 °C/1013 hPa

mp

-10 °C

transition temp

flash point 86 °C

solubility

<1 g/L

density

1.121-1.127 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H7NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H

InChI key

NNICRUQPODTGRU-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 1.121 - 1.127
Identity (IR): passes test
Color (visual): yellow to brown

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service