Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
[(C6H5)3P]2PdCl2
CAS Number:
Molecular Weight:
701.90
UNSPSC Code:
12352103
EC Index Number:
237-744-2
NACRES:
NA.22
MDL number:
Product Name
Bis(triphenylphosphine)palladium(II) chloride (15.2% Pd), for synthesis
Quality Level
form
crystals
mp
>300 °C
storage temp.
2-30°C
SMILES string
[Pd]([P](c6ccccc6)(c5ccccc5)c4ccccc4)([P](c3ccccc3)(c2ccccc2)c1ccccc1)(Cl)Cl
InChI
1S/2C18H15P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H;2*1H;/q;;;;+2/p-2
InChI key
YNHIGQDRGKUECZ-UHFFFAOYSA-L
Analysis Note
Assay (to Pd): ≥ 99.0 %
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 4 - Skin Sens. 1A
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8041740001 | 04022536384153 |
| 8041748500 | 04027269278731 |
| 8041740005 | 04027269070694 |
