Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18899

Sigma-Aldrich

Sebacoyl dichloride

for synthesis

Synonym(s):

Sebacoyl dichloride, Sebacoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClOC(CH2)8COCl
CAS Number:
Molecular Weight:
239.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-843-4
NACRES:
NA.22

Quality Level

form

liquid

potency

400 mg/kg LD50, oral (Rat)

bp

161 °C/19 hPa

mp

-5 °C

transition temp

flash point 113 °C

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)CCCCCCCCC(=O)Cl

InChI

1S/C10H16Cl2O2/c11-9(13)7-5-3-1-2-4-6-8-10(12)14/h1-8H2

InChI key

WMPOZLHMGVKUEJ-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 99.0 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service