Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.18531

Sigma-Aldrich

(2R,3R)-(+)-Tartaric acid

for resolution of racemates for synthesis

Synonym(s):

(2R,3R)-(+)-Tartaric acid, 2,3-Dihydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(OH)CH(OH)COOH
CAS Number:
Molecular Weight:
150.09
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-766-0
NACRES:
NA.22

grade

for synthesis

Quality Level

vapor pressure

<5 Pa ( 20 °C)

Assay

≥99% (acidimetric)

form

solid

autoignition temp.

425 °C

pH

1.6 (25 °C, 100 g/L in H2O)

mp

168-170 °C

transition temp

flash point 210 °C

solubility

1390 g/L

density

1.76 g/cm3 at 20 °C

bulk density

1000 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)C(O)C(O)C(=O)[O-].[H+].[H+]

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)

InChI key

FEWJPZIEWOKRBE-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Spec. rotation [α²0/D (c=20 in water): +12.0 - +13.0 °
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service