Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18449

Sigma-Aldrich

4,4′-Thiobis(2-tert-butyl-5-methylphenol)

for synthesis

Synonym(s):

4,4′-Thiobis(2-tert-butyl-5-methylphenol), Bis(3-tert-butyl-4-hydroxy-6-methylphenyl)sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(2-(CH3)-4-(OH)-5-((CH3)3C)C6H2)2S
CAS Number:
Molecular Weight:
358.54
UNSPSC Code:
12352104
EC Index Number:
202-525-2
NACRES:
NA.22

Quality Level

Assay

≥99.0% (HPLC)

form

powder

autoignition temp.

402 °C

potency

2345 mg/kg LD50, oral (Rat)

mp

158-162 °C

transition temp

flash point 215 °C

storage temp.

2-30°C

InChI

1S/C22H30O2S/c1-13-9-17(23)15(21(3,4)5)11-19(13)25-20-12-16(22(6,7)8)18(24)10-14(20)2/h9-12,23-24H,1-8H3

InChI key

HXIQYSLFEXIOAV-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 158 °C
Melting range (upper value): ≤ 162 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service