Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18419

Sigma-Aldrich

3-Fluorobenzyl chloride

for synthesis

Synonym(s):

3-Fluorobenzyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClF
CAS Number:
Molecular Weight:
144.57
MDL number:
UNSPSC Code:
12352101
EC Index Number:
207-264-8
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

bp

176-177 °C/1013 hPa

transition temp

flash point 58 °C

density

1.21 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H6ClF/c8-5-6-2-1-3-7(9)4-6/h1-4H,5H2

InChI key

XBDXMDVEZLOGMC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.212 - 1.214
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service