Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F7601

Sigma-Aldrich

2-Fluorobenzyl chloride

98%

Synonym(s):

α-Chloro-2-fluorotoluene, 1-Chloromethyl-2-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2Cl
CAS Number:
Molecular Weight:
144.57
Beilstein:
471699
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.514 (lit.)

bp

86 °C/40 mmHg (lit.)

density

1.216 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1CCl

InChI

1S/C7H6ClF/c8-5-6-3-1-2-4-7(6)9/h1-4H,5H2

InChI key

MOBRMRJUKNQBMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service