Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14785

Sigma-Aldrich

Isopropyl 2-bromoisobutyrate

for synthesis

Synonym(s):

Isopropyl 2-bromoisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13BrO2
CAS Number:
Molecular Weight:
209.08
MDL number:
UNSPSC Code:
12352108
EC Index Number:
257-158-0
NACRES:
NA.22

Quality Level

form

liquid

transition temp

flash point 63 °C

density

1.24 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrC(C)(C)C(=O)OC(C)C

InChI

1S/C7H13BrO2/c1-5(2)10-6(9)7(3,4)8/h5H,1-4H3

InChI key

UNZJYKKJZGIFCG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

145.4 °F

Flash Point(C)

63 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service