Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.14759

Sigma-Aldrich

Terpineol

(mixture of isomers) for synthesis

Synonym(s):

Terpineol isomer mixture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
232-268-1
NACRES:
NA.22

vapor pressure

3 hPa ( 20 °C)

Quality Level

Assay

≥80% (GC)

form

solid or liquid

potency

4300 mg/kg LD50, oral (Rat)

bp

213-218 °C (lit.)

mp

15-35 °C

transition temp

flash point 88 °C

density

0.934 g/mL at 20 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Encapsulation and release of 4-terpineol: A study described the encapsulation of 4-terpineol with β-cyclodextrin, detailing the inclusion mechanism and characterization, along with a humidity-triggered release system. This highlights its potential in controlled delivery applications (Cheng et al., 2024).

Analysis Note

Assay isomers (GC, area%): ≥ 80.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.932 - 0.936
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service