Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

30627

Supelco

α-Terpineol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2325137
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

composition

γ-Terpineol, ≤10.0% GC (minor component)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.480-1.486
n20/D 1.482 (lit.)

bp

217-218 °C (lit.)

mp

31-35 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1=CC[C@H](CC1)C(C)(C)O

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3

InChI key

WUOACPNHFRMFPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity of Melaleuca alternifolia (tea tree) essential oil.[1] The reaction rate constant of α-terpineol with the OH radical and ozone was found to be (1.9±0.5)×10-10cm3 molecule-1s-1 and (3.0±0.2)×10-16cm3 molecule-1s-1, respectively.[2]

Application

α-Terpineol has been used as reference standard in GC-MS analysis for the detection of terpenoids extracted by hexane.[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum mentha pimpinella salvia thymus valeriana glycyrrhiza

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Releasing potential power of terpene synthases by a robust precursor supply platform
Bian, Guangkai, et al.
Metabolic engineering, 42, 1-8 (2017)
Antifungal activity of the components of Melaleuca alternifolia (tea tree) oil.
Hammer KA , et al.
Journal of Applied Microbiology, 95(4), 853-860 (2003)
J R Wells
Environmental science & technology, 39(18), 6937-6943 (2005-10-06)
A bimolecular rate constant, kOH+alpha-terpineol, of (1.9 +/- 0.5) x 10(-10) cm3 molecule(-1) s(-1) was measured using gas chromatography/mass spectrometry and the relative rate technique for the reaction of the hydroxyl radical (OH) with alpha-terpineol (1-methyl-4-isopropyl-1-cyclohexen-8-ol) at (297 +/- 3)
Changkyu Kim et al.
Nanoscale, 7(15), 6627-6635 (2015-03-21)
The realization of air-stable nanoparticles, well-formulated nanoinks, and conductive patterns based on copper is a great challenge in low-cost and large-area flexible printed electronics. This work reports the synthesis of a conductively interconnected copper structure via thermal sintering of copper
Stuart J Lansdell et al.
Molecular pharmacology, 87(1), 87-95 (2014-10-24)
In common with other members of the Cys-loop family of pentameric ligand-gated ion channels, 5-hydroxytryptamine type 3 receptors (5-HT3Rs) are activated by the binding of a neurotransmitter to an extracellular orthosteric site, located at the interface of two adjacent receptor

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service