Skip to Content
MilliporeSigma

8.14656

Antimony(III) chloride

for synthesis

Synonym(s):

Antimony(III) chloride, Antimonous acid, Antimony trichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Cl3Sb
CAS Number:
Molecular Weight:
228.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-047-2
NACRES:
NA.22
Grade:
synthesis grade
Form:
solid
Solubility:
931 g/L (Hydrolysis)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

0.16 hPa ( 20 °C)

form

solid

bp

233 °C/1013 hPa

mp

73 °C

solubility

931 g/L (Hydrolysis)

density

3.14 g/cm3 at 20 °C

bulk density

1600 kg/m3

storage temp.

2-30°C

InChI

1S/C22H23ClN2O8.3ClH.Sb/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;;;;/h4-5,7-8,15,26-27,31-33H,6,24H2,1-3H3;3*1H;/q;;;;+3/p-3/b20-13+;;;;/t7-,8-,15-,21-,22-;;;;/m0..../s1

InChI key

LIARQHHWPMTMFO-CRPMINNHSA-K

Application

  • The metal halide structure and the extent of distortion control the photo-physical properties of luminescent zero dimensional organic-antimony (iii) halide hybrids: Relevant to material scientists, this research discusses the synthesis of organic-antimony halide hybrids for use in luminescent materials (Biswas et al., 2021).
  • A deep-red-emission antimony (iii) chloride with dual-cations: This study is particularly relevant to material science, detailing the synthesis and luminescent properties of a novel antimony(III) chloride complex, highlighting its potential in optoelectronic applications (Peng et al., 2021).
  • Antimony (III) halide compounds of thioureas: Structures and biological activity: This article focuses on the biological activities of antimony(III) halide complexes, pertinent to medicinal chemists interested in novel therapeutic agents (Han et al., 2014).

Analysis Note

Assay (iodometric): ≥ 99.0 %
Identity (ICP-OES): passes test


Still not finding the right product?

Explore all of our products under Antimony(III) chloride


pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library