Skip to Content
MilliporeSigma
Antimony(III) chloride JIS special grade, ≥98.0%

Antimony(III) chloride

Antimony(III) chloride JIS special grade, ≥98.0%
Synonym(s):
Antimony trichloride, Antimony(III) chloride
Linear Formula:
SbCl3
CAS Number:
Molecular Weight:
228.12
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade