Skip to Content
MilliporeSigma
All Photos(2)

Documents

8.14576

Sigma-Aldrich

Bis(2-ethylhexyl) sebacate

for synthesis

Synonym(s):

Bis(2-ethylhexyl) sebacate, Dioctyl sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H50O4
CAS Number:
Molecular Weight:
426.67
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-558-8
NACRES:
NA.22

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

>250 °C/1013 hPa

mp

-55 °C

transition temp

flash point >200 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C26H50O4/c1-5-9-17-23(7-3)21-29-25(27)19-15-13-11-12-14-16-20-26(28)30-22-24(8-4)18-10-6-2/h23-24H,5-22H2,1-4H3

InChI key

VJHINFRRDQUWOJ-UHFFFAOYSA-N

Application

  • Optimization of ion-selective sensors: Research highlights the critical role of Bis(2-ethylhexyl) sebacate in enhancing the performance of ion-selective sensors, particularly by optimizing the combination of ion-exchangers and plasticizers to improve sensor responsiveness and durability (Stelmach et al., 2024).
  • Lubricant viscosity under variable temperatures: A study examining the viscosity of ester-based lubricants, including Bis(2-ethylhexyl) sebacate, across different temperatures, provided insights essential for industrial applications requiring consistent performance under varying thermal conditions (Sarpa et al., 2023).
  • Study of ionic liquid behavior in plasticized membranes: Research involving Bis(2-ethylhexyl) sebacate explored its effects on the properties of polymeric membranes used with ionic liquids, contributing to a better understanding of membrane behaviors in electrochemical applications (Pokhvishcheva et al., 2022).
  • Improvement of biodegradable plastics: The application of Bis(2-ethylhexyl) sebacate in poly(lactic acid) for the packaging industry was investigated, demonstrating its effectiveness in enhancing the flexibility and processability of biodegradable plastics (Gzyra-Jagieła et al., 2021).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.913 - 0.916
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

awg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service