Skip to Content
MilliporeSigma

1.00263

Formic acid

98-100%, EMPROVE® ESSENTIAL

Synonym(s):

Formic acid, Methanoic acid, Formylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
EC Index Number:
200-579-1
UNSPSC Code:
12352106
Beilstein/REAXYS Number:
1209246
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

1.6 (vs air)

Quality Segment

vapor pressure

44.8 mmHg ( 20 °C)

product line

EMPROVE® ESSENTIAL

assay

≥98% (alkalimetric)

form

liquid

autoignition temp.

1004 °F

expl. lim.

12-38 % (v/v), 57 %

concentration

98-100%

technique(s)

API processing | polymorph screening: suitable, API processing | salt formation: suitable

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 10 g/L in H2O)

bp

100-101 °C (lit.), 101 °C/1013 hPa

mp

8.2-8.4 °C (lit.)

transition temp

flash point 49.5 °C

density

1.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%, sulfate (SO42-): ≤0.004%, sulfite (SO32-): ≤0.002%

cation traces

As: ≤0.0003%, Cu: ≤0.002%, Pb: ≤0.001%, Zn: ≤0.002%, heavy metals (as Pb): ≤0.0005%

application(s)

liquid formulation
pharmaceutical

storage temp.

15-25°C

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Formic acid can be used as acidifying agent and buffer component in a variety of pharmaceutical formulations and processes.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Formic acid


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

supp_hazards

Storage Class

3 - Flammable liquids

flash_point_f

116.6 °F

flash_point_c

47 °C



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library