Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.10298

Sigma-Aldrich

Benzyltriethylammonium chloride

for synthesis

Synonym(s):

Benzyltriethylammonium chloride, Triethylbenzylammonium chloride, BTEAC, TEBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2(C2H5)3NCl
CAS Number:
Molecular Weight:
227.77
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-270-1
NACRES:
NA.22

Quality Level

autoignition temp.

300 °C

potency

2219 mg/kg LD50, oral (Rat)

pH

6-8 (20 °C, 100 g/L in H2O)

mp

190-192 °C (decomposition)

transition temp

flash point >275 °C

solubility

630 g/L

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[N+](CC)(CC)(CC)Cc1ccccc1

InChI

1S/C13H22N.ClH/c1-4-14(5-2,6-3)12-13-10-8-7-9-11-13;/h7-11H,4-6,12H2,1-3H3;1H/q+1;/p-1

InChI key

HTZCNXWZYVXIMZ-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

527.0 °F - closed cup

Flash Point(C)

> 275 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service