Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.06548

Sigma-Aldrich

3-Nitrobenzenesulfonic acid sodium salt

for synthesis

Synonym(s):

3-Nitrobenzenesulfonic acid sodium salt, Sodium 3-nitrobenzenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(NO2)C6H4SO3Na
CAS Number:
Molecular Weight:
225.15
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-857-3
NACRES:
NA.22
form:
needles

Quality Level

form

needles

autoignition temp.

355 °C

potency

>2000 mg/kg LD50, oral (Rat)

pH

8 (23 °C, 50 g/L in H2O)

mp

350 °C (decomposition)

solubility

200 g/L

density

0.45 g/cm3 at 20 °C

bulk density

450 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])c1cc(ccc1)[N+](=O)[O-]

InChI

1S/C6H5NO5S.Na/c8-7(9)5-2-1-3-6(4-5)13(10,11)12;/h1-4H,(H,10,11,12);/q;+1/p-1

InChI key

LJRGBERXYNQPJI-UHFFFAOYSA-M

Analysis Note

Assay (acidimetric after ion exchange): ≥ 92.0 %
Water (K. F.): ≤ 5.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service