Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07490

Sigma-Aldrich

Polyethylene glycol

4000, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 4000, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 4000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

>320 °C (DIN 51794)

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

53-58 °C

transition temp

flash point 270 °C

solubility

500 g/L

density

1.2 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 4000 for synthesis. CAS No. 25322-68-3, EC Number 500-038-2.

Analysis Note

Hydroxyl value: 25 - 32
Melting range (lower value): ≥ 58 °C
Melting range (upper value): ≤ 62 °C
Average molecular mass: 3500 - 4500
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service