Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.03282

Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene

for synthesis

Synonym(s):

1,8-Diazabicyclo[5.4.0]undec-7-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2
Molecular Weight:
152.24
MDL number:
UNSPSC Code:
12352005
EC Index Number:
229-713-7
NACRES:
NA.22

vapor pressure

0.02 hPa ( 25 °C)

Quality Level

form

liquid

autoignition temp.

260 °C

potency

215-681 mg/kg LD50, oral (Rat)

expl. lim.

1.1-6.5 % (v/v)

pH

12.8 (20 °C, 10 g/L in H2O)

kinematic viscosity

5.48 cSt(40 °C)

mp

<-70 °C

transition temp

flash point 116 °C

solubility

soluble 1000 g/L

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H16N2/c1-2-5-9-10-6-4-8-11(9)7-3-1/h1-8H2

InChI key

GQHTUMJGOHRCHB-UHFFFAOYSA-N

Application

  • Depolymerization of poly (bisphenol A carbonate) under mild conditions by solvent-free alcoholysis catalyzed by 1, 8-diazabicyclo undec-7-ene as a recyclable: DBU catalyzes the depolymerization of polycarbonates under mild, solvent-free conditions, demonstrating its effectiveness and recyclability as a catalyst (E Quaranta, D Sgherza, G Tartaro, 2017).
  • Activating cellulose via its reversible reaction with CO2 in the presence of 1, 8-diazabicyclo undec-7-ene for the efficient synthesis of cellulose acetate: This research shows how DBU can activate cellulose for a reversible reaction with CO2, facilitating the synthesis of cellulose acetate (Y Yang, L Song, C Peng, E Liu, H Xie, 2015).
  • Bifunctional 1, 8-Diazabicyclo undec-7-ene for Visible Light-Induced Heck-Type Perfluoroalkylation of Alkenes: DBU acts as a bifunctional organocatalyst for visible light-induced Heck-type perfluoroalkylation of alkenes, showcasing its potential in photoredox catalysis (L Tang, G Lv, Y Fu, XP Chang, R Cheng, 2022).
  • Cooperative calcium-based catalysis with 1, 8-diazabicyclo undec-7-ene for the cycloaddition of epoxides with CO2 at atmospheric pressure: This study demonstrates the cooperative catalysis of CaBr2 and DBU in the cycloaddition of epoxides with CO2, highlighting a sustainable approach to carbonate synthesis (X Liu, S Zhang, QW Song, XF Liu, R Ma, LN He, 2016).

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F

Flash Point(C)

116 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service