Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

595128

Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene, polymer-bound

100-200 mesh, extent of labeling: 1.5-2.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

1,8-Diazabicyclo[5.4.0]undec-7-ene on Polystyrene, DBU on PS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

1.5-2.5 mmol/g loading

particle size

100-200 mesh

Application

Polymer-supported base

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamura, Y.
Synthetic Communications, 24, 2907-2907 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service