Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02936

Sigma-Aldrich

2,2-Dimethoxypropane

for synthesis

Synonym(s):

2,2-Dimethoxypropane, Acetone dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O2
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352112
EC Index Number:
201-056-0
NACRES:
NA.22
grade:
for synthesis
vapor pressure:
60 hPa ( 16 °C)

grade

for synthesis

Quality Level

vapor pressure

60 hPa ( 16 °C)

Assay

≥97.0% (GC)

form

liquid

potency

>2260 mg/kg LD50, oral (Rat)
>2100 mg/kg LD50, skin (Rat)

expl. lim.

6-31 % (v/v)

dilution

(for synthesis)

mp

-47 °C

transition temp

flash point -10 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(OC)(C)C)C

InChI

1S/C5H12O2/c1-5(2,6-3)7-4/h1-4H3

InChI key

HEWZVZIVELJPQZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.850 - 0.851
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F

Flash Point(C)

-10 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service