Skip to Content
MilliporeSigma
All Photos(1)

Documents

7710-OP

Millipore

Sodium Chloride

OmniPur® Grade, >=99.5%

Synonym(s):

Sodium chloride, NaCl, Halite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

Quality Level

form

granular (Fine)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

801 °C (lit.)

shipped in

ambient

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Biology Grade.

A260 (0.1 M, water): ≤0.01 AU
A280 (0.1 M, water): ≤0.01 AU
Assay: ≥99.5%
Barium (Ba): ≤0.001%
Calcium, magnesium, and R2O3 precipitate: ≤0.005%
DNAse: none detected
Heavy metals (as Pb): ≤0.0002%
Insoluble matter: ≤0.005%
Iodide (I): ≤0.002%
pH of a 5% solution at 25°C: 5.5-7.5
Phosphatase: none detected
Phosphate (PO4): ≤0.0005%
Potassium (K): ≤0.005%
Protease: none detected
RNAse: none detected
Sulfate (SO4): ≤0.003%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service