Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.06406

Supelco

Sodium chloride

99.99 Suprapur®

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-598-3
NACRES:
NB.21
Assay:
99.99%
grade:
for inorganic trace analysis
form:
solid
solubility:
358 g/L

grade

for inorganic trace analysis

Quality Level

vapor pressure

1.3 hPa ( 865 °C)

Assay

99.99%

form

solid

potency

3000 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C/1013 hPa

mp

801 °C (lit.)

solubility

358 g/L

bulk density

1140 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

99.99 Suprapur®

Application

Sodium chloride 99.99 Suprapur®. CAS No. 7647-14-5, EC Number 231-598-3.

Linkage

Replaces: 6406; 6406-2; 6406-1

Analysis Note

Purity (metallic)min 99.99 %Assay (argentometric)min 99.5 %Bromide (Br)max 10 ppmIodide (I)max 10 ppmPhosphate (PO₄)max 5 ppmSulphate (SO₄)max 10 ppmTotal nitrogen (N)max 10 ppmHexacyanoferrate (Fe(CN)₆)max 1 ppmAl (Aluminium)max 0.01 ppmBa (Barium)max 2 ppmCa (Calcium)max 0.1 ppmCd (Cadmium)max 0.005 ppmCe (Cerium)max 0.005 ppmCo (Cobalt)max 0.005 ppmCr (Chromium)max 0.01 ppmCs (Cesium)max 5 ppmCu (Copper)max 0.005 ppmEu (Europium)max 0.005 ppmFe (Iron)max 0.05 ppmK (Potassium)max 5 ppmLa (Lanthanum)max 0.005 ppmLi (Lithium)max 0.5 ppmMg (Magnesium)max 0.05 ppmMn (Manganese)max 0.01 ppmNi (Nickel)max 0.01 ppmPb (Lead)max 0.005 ppmRb (Rubidium)max 0.5 ppmSc (Scandium)max 0.005 ppmSm (Samarium)max 0.005 ppmSr (Strontium)max 0.1 ppmTl (Thallium)max 0.005 ppmY (Yttrium)max 0.005 ppmYb (Ytterbium)max 0.005 ppmZn (Zinc)max 0.01 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Preparation of a standard solution for COD/chloride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service