Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6910-OP

Millipore

PIPES Sodium Salt

OmniPur® Grade, >=99.0%

Synonym(s):

OmniPur® PIPES, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H33N4Na3O12S4
CAS Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Quality Level

form

crystalline powder (Fine)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

[Na+].[Na+].[Na+].[S](=O)(=O)([O-])CCN2CCN(CC2)CC[S](=O)(=O)O.[S](=O)(=O)([O-])CCN1CCN(CC1)CC[S](=O)(=O)[O-]

InChI

1S/2C8H18N2O6S2.3Na/c2*11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;;/h2*1-8H2,(H,11,12,13)(H,14,15,16);;;/q;;3*+1/p-3

InChI key

FPDACHMSOMILQI-UHFFFAOYSA-K

General description

Molecular biology grade.

Specifications:
Assay (anhydrous basis): ≥99.0%
DNase: None detected
Heavy metals (as Pb): ≤0.0001%
pKa at 25°C 6.10-7.50
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service