Skip to Content
MilliporeSigma

P3768

PIPES disodium salt

≥99% (titration)

Synonym(s):

1,4-Piperazinediethanesulfonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) disodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H16N2Na2O6S2
CAS Number:
Molecular Weight:
346.33
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
278-562-3
Beilstein/REAXYS Number:
9453872
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (titration)

form

crystalline powder

useful pH range

6.1-7.5

pKa (25 °C)

6.8

solubility

water: 0.5 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O6S2.2Na/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;/h1-8H2,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

GMHSTJRPSVFLMT-UHFFFAOYSA-L



Still not finding the right product?

Explore all of our products under PIPES disodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library