Skip to Content
MilliporeSigma

684274

β1,2-Xylosidase, Xanthomonas sp.

Synonym(s):

β1,2-Xylosidase, Xanthomonas sp., β-D-Xylanxylohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352202
NACRES:
NA.54
MDL number:
Specific activity:
≥10 units/mL, ≥20 units/mg protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

specific activity

≥10 units/mL, ≥20 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

β-N-acetylglucosaminidase, α-fucosidase, α- and β-galactosidase, α-mannosidase, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Native β1,2-Xylosidase from Xanthomonas species. Catalyzes the hydrolysis of terminal xylose β1,2-linked to the β-mannose of N-linked oligosaccharides. Enzyme requires removal of α1,3-linked mannose. Enzyme requires 5 mM calcium chloride in reaction buffer for full activity.
Note: 1 mU = 1 milliunit.

Physical form

In 25 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Preparation Note

Following initial use, refrigerate (4°C).

Other Notes

One unit is defined as the amount of enzyme that will release 1 nmol of methylumbelliferone from 4-methylumbelliferyl-7-β-D-xyloside per min at 37°C, pH 5.0.
Reese, A.J. and Doening, T.L. 2003. Mol. Microbiol.50, 1401.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library