Skip to Content
MilliporeSigma
All Photos(1)

Documents

670111-M

Millipore

UDP-α-D-Galactose, Disodium Salt

A donor substrate for galactosyltransferases used in the biosynthesis of galactose-containing oligosaccharides. Decreased red blood cell levels of UDP-Gal are found in patients with maple syrup urine disease or phenylketonuria.

Synonym(s):

UDP-α-D-Galactose, Disodium Salt, UDP-Gal, Uridine-5ʹ-diphosphogalactose, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O17P2 · 2Na
CAS Number:
Molecular Weight:
610.27
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.51

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 100 mg/mL

storage temp.

−20°C

InChI

1S/C15H24N2O17P2.2Na/c18-3-5-8(20)10(22)12(24)14(32-5)33-36(28,29)34-35(26,27)30-4-6-9(21)11(23)13(31-6)17-2-1-7(19)16-15(17)25;;/h1-2,5-6,8-14,18,20-24H,3-4H2,(H,26,27)(H,28,29)(H,16,19,25);;/q;2*+1/p-2/t5-,6-,8+,9-,10+,11-,12-,13-,14-;;/m1../s1

InChI key

PKJQEQVCYGYYMM-QKYKBPIOSA-L

General description

A donor substrate for galactosyltransferases used in the biosynthesis of galactose-containing oligosaccharides. Decreased red blood cell levels of UDP-Gal are found in patients with maple syrup urine disease or phenylketonuria.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Stults, C.L., et al. 1999. Glycobiology 9, 661.
Gibson, J.B., et al. 1996. Am. J. Clin. Nutr. 63, 704.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service