Skip to Content
MilliporeSigma

670107

UDP-α-D-N-Acetylglucosamine, Disodium Salt

Donor substrate for N-acetylglucosaminyltransferases. Useful in the synthesis of aryl azide derivatives that can be used in the affinity labeling of glycosyltransferase and UDP-HexNAc pyrophosphorylase.

Synonym(s):

UDP-α-D-N-Acetylglucosamine, Disodium Salt, UDP-GlcNAc, 2Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H25N3O17P2 · 2Na
CAS Number:
Molecular Weight:
651.32
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Form:
crystalline powder
Assay:
≥95% (HPLC)
Solubility:
water: 50 mg/mL
Color:
white to off-white
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])(O[P](=O)([O-])OC[C@H]2O[C@H]([C@@H]([C@@H]2O)O)[n]3[c]([nH][c](cc3)=O)=O)O[C@H]1O[C@@H]([C@H]([C@@H]([C@H]1NC(=O)C)O)O)CO

InChI

1S/C17H27N3O17P2.2Na/c1-6(22)18-10-13(26)11(24)7(4-21)35-16(10)36-39(31,32)37-38(29,30)33-5-8-12(25)14(27)15(34-8)20-3-2-9(23)19-17(20)28;;/h2-3,7-8,10-16,21,24-27H,4-5H2,1H3,(H,18,22)(H,29,30)(H,31,32)(H,19,23,28);;/q;2*+1/p-2/t7-,8-,10-,11-,12-,13-,14-,15-,16-;;/m1../s1

InChI key

HXWKMJZFIJNGES-YZVFIFBQSA-L

General description

Donor substrate for N-acetylglucosaminyltransferases. Useful in the synthesis of aryl azide derivatives that can be used in the affinity labeling of glycosyltransferase and UDP-HexNAc pyrophosphorylase.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zeng, Y., et al. 1996. Anal. Biochem. 239, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library