Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6630

Millipore

Uracil

Synonym(s):

Uracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

description

RTECS - YQ8650000
Merck USA index - 14, 9850

Quality Level

Assay

≥99% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to slightly yellowish

solubility

hot water: 1 mg/mL

cation traces

heavy metals (as Pb): ≤10 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

n1c(nccc1O)O

InChI

1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ISAKRJDGNUQOIC-UHFFFAOYSA-N

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service