Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648313

Millipore

TRIS Hydrochloride

ULTROL® Grade, ≥99% (titration), useful in pH range 7.0-9.0

Synonym(s):

Trizma® hydrochloride, tris(Hydroxymethyl)aminomethane, HCl, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.25

description

Merck USA index - 14, 9772

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

7.0-9.0

pKa (25 °C)

8.1

solubility

water: 500 mg/mL

cation traces

heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Absorbance (1.0 M, H2O, 260 nm):≤0.1.
Useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.
Widely used as a biological buffer (absorbance: <0.1 at 260 nm, 1.0 M, H2O). A 1.0 M solution in H2O has a pH range of 3.5-5.0 and a pKa of 8.3 at 20°C.

Application


  • Effect of Organic Solvents on the Activity, Stability and Secondary Structure of asclepain cI, Using FTIR and Molecular Dynamics Simulations.: This study uses Tris hydrochloride as a buffer to assess the stability and structural changes in the asclepain cI enzyme, providing insights into enzyme applications in industrial and medical biotechnology (Origone et al., 2024).

  • Microencapsulation of Omeprazole by Lactobacillus acidophilus ATCC 4356 Surface Layer Protein and Evaluation of its Stability in Acidic Condition.: Research demonstrates the use of Tris hydrochloride buffer in stabilizing microencapsulated omeprazole, aimed at enhancing drug delivery in acidic environments (Mobarak Qamsari et al., 2020).

Warning

Toxicity: Standard Handling (A)

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiulan Shao et al.
Oncology letters, 21(6), 431-431 (2021-04-20)
Ovarian cancer is one of the leading causes of cancer-related death among women worldwide and accounts for 4% of all cancer cases in female patients. To date, ovarian cancer has the poorest prognosis among all types of gynecological cancer; thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service