Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616424

Sigma-Aldrich

Transferrin, Holo, Human Plasma, Low Endotoxin

Synonym(s):

Transferrin, Holo, Human Plasma, Low Endotoxin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

description

Merck USA index - 14, 9571

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

pI 

5.8

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native holotransferrin from human plasma. Transferrin is a major iron binding protein and is found in human plasma or serum at concentrations of 3 mg/ml. Its concentration correlates with the total iron binding capacity of serum and its level is regulated by the availability of iron. It has bacteriostatic and fungistatic activity and is essential for iron utilization by the hemin-synthesizing reticulocytes. Stored in bone marrow as Tf-bound iron. pI: 5.8.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 150 mM NaCl, 20 mM sodium phosphate buffer, pH 7.4.

Preparation Note

Please refer to the vial label for the pre-lyophilization concentration.
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or add NaN₃ and refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 2 weeks at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service