Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

613548

Sigma-Aldrich

TMB, Insoluble

Synonym(s):

TMB, Insoluble, 3,3ʹ,5,5ʹ-Tetramethylbenzidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.41

form

liquid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clearlight brown

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3

InChI key

UAIUNKRWKOVEES-UHFFFAOYSA-N

General description

Insoluble TMB substrate supplied as a single component system. TMB is the most sensitive chromogenic substrate for detection of horseradish peroxidase (HRP)-labeled probes. In the presence of HRP and hydrogen peroxide (H2O2), TMB is oxidized first to a blue, cation free radical; upon further reaction with HRP/H2O2, or addition of acid, the radical is converted to a yellow diimine terminal oxidation product. TMB is used routinely in ELISA procedures, whereby its soluble oxidation products are measured spectrophotometrically. For TMB to be worthwhile as a blotting reagent, enhancers such as dextran sulfate or other sugar polymers are usually added.This TMB reagent is a safe, single component system that has been developed for use in blotting procedures by a proprietary methodology that eliminates the use of sugar polymers. The reagent produces a stable, insoluble aquamarine precipitate at the reaction site with little or no background. It can significantly increase the detection limits of assays on a variety of membranes. Ideal for many immunoblotting procedures and can significantly increase detection limits of assays on a variety of membranes
Peroxidase substrate that produces a stable, insoluble, aquamarine blue precipitate at reaction site with little or no background.

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Physical form

1.19 mM H₂O₂ in a proprietary solvent, 1.13 mM TMB, pH 4.9 ± 0.03.

Reconstitution

Discard if the solution turns blue or turbid. The reagent is stable at room temperature; refrigeration is recommended for long-term storage. Warm to assay temperature before use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Western blotting involves protein separation by gel electrophoresis, protein transfer to a polyvinylidene difluoride (PVDF) or nitrocellulose membrane, and detection by various methods.

Western blotting involves protein separation by gel electrophoresis, protein transfer to a polyvinylidene difluoride (PVDF) or nitrocellulose membrane, and detection by various methods.

Western blotting involves protein separation by gel electrophoresis, protein transfer to a polyvinylidene difluoride (PVDF) or nitrocellulose membrane, and detection by various methods.

Western blotting involves protein separation by gel electrophoresis, protein transfer to a polyvinylidene difluoride (PVDF) or nitrocellulose membrane, and detection by various methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service