Skip to Content
MilliporeSigma
All Photos(4)

Documents

6003

Millipore

Amicon® Stirred Cell Selector Valve

The selector valve provides a quick and simple way to switch the Amicon Stirred Cell between concentration and diafiltration modes without interrupting system operation.

Synonym(s):

Selector valve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
32019190
NACRES:
NB.24

material

Nylon (chemically sanitizable)
nitrile (chemically sanitizable)
polyacetal (chemically sanitizable)

Quality Level

L × W × H

7.5 cm × 2 cm × 8 cm

application(s)

sample preparation
sample preparation

shipped in

ambient

General description

Valve with sliding control for instant switching from concentration to diafiltration, or switching gas and liquid lines simultaneously. Simplifies operation and avoids the need for multiple T-fittings and valves.

Application

Four tubing connectors lead to the gas supply, the stirred cell, and the inlet and outlet ports of a reservoir (e.g. Amicon Stirred Cell Reservoir, cat no 6028).

Other Notes

7.5 x 2 x 8 cm

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Protocols

Demonstrating the effective use of Amicon Stirred Cell for large volume concentration through diafiltration.

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service