Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.89588

Sigma-Aldrich

o-Xylene

97%, anhydrous

Synonym(s):

1,2-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1815558
MDL number:
UNSPSC Code:
12191501
NACRES:
NA.22
grade:
anhydrous
bp:
143-145 °C (lit.)
vapor pressure:
<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

3.7 (vs air)

vapor pressure

<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

Assay

97%

autoignition temp.

867 °F

expl. lim.

7 %

refractive index

n20/D 1.505 (lit.)

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

solubility

water: partially soluble 0.1705 g/L at 25 °C

density

0.879 g/mL at 20 °C (lit.)

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service