Skip to Content
MilliporeSigma
All Photos(1)

Documents

580549

Millipore

Tauroursodeoxycholic Acid, Sodium Salt

Tauroursodeoxycholic is a detergent useful for the solubilization of lipids and membrane-bound proteins.

Synonym(s):

Tauroursodeoxycholic Acid, Sodium Salt, Sodium Tauroursodeoxycholate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NO6S · Na
CAS Number:
Molecular Weight:
521.69
UNSPSC Code:
12352106
NACRES:
NA.21

Quality Level

Assay

≥97% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C26H45NO6S/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33)/t16-,17+,18-,19-,20?,21?,22+,24?,25+,26-/m1/s1

InChI key

BHTRKEVKTKCXOH-AYSJQVDDSA-N

General description

Detergent useful for the solubilization of lipids and membrane–bound proteins.
Detergent useful for the solubilization of lipids and membrane-bound proteins. Has cryoprotective effects against hydrophobic bile salts in rats. Also reported to inhibit pepsin activity in vitro.

Application


  • Inhibition of endoplasmic reticulum stress combined with activation of angiotensin-converting enzyme 2: novel approach for the prevention of endothelial dysfunction in type 1 diabetic rats.: This study highlights the efficacy of tauroursodeoxycholic acid, sodium salt in reducing endoplasmic reticulum stress, suggesting its utility in preventing vascular complications in diabetes (Gaikwad AB et al., 2022).

Warning

Toxicity: Standard Handling (A)

Other Notes

Ozcan, U., et al. 2006. Science313, 1137.
Miyake, H., et al. 1999. Dig. Dis. Sci.44, 197.
Eto, T. and Tompkins, R.K. 1985. Am. J. Surg.150, 564.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Xie et al.
The Journal of biological chemistry, 282(45), 33043-33051 (2007-09-15)
Mammalian small intestinal lipid absorption requires the coordinated interactions of apolipoprotein B (apoB) and the microsomal triglyceride transfer protein (Mttp). The observation that apoB100 displays greater dependence on Mttp availability than does apoB48 prompted us to examine the phenotype of
Maddalena Parafati et al.
Disease models & mechanisms, 11(9) (2018-09-27)
Hepatic steatosis, a reversible state of metabolic dysregulation, can promote the onset of nonalcoholic steatohepatitis (NASH), and its transition is thought to be critical in disease evolution. The association between endoplasmic reticulum (ER) stress response and hepatocyte metabolism disorders prompted
Isabel Castro et al.
Rheumatology (Oxford, England), 59(4), 742-753 (2019-08-05)
Xerostomia in SS patients has been associated with low quality and quantity of salivary mucins, which are fundamental for the hydration and protection of the oral mucosa. The aim of this study was to evaluate if cytokines induce aberrant mucin
Huika Xia et al.
Investigative ophthalmology & visual science, 56(11), 6638-6645 (2015-10-16)
To investigate the effects of tauroursodeoxycholic acid (TUDCA) and alpha-lipoic-acid (ALA) on the visual response properties of cat retinal ganglion cells (RGCs) in wholemount retinas. Young adult cats were divided into three groups: control, ALA, and TUDCA. In vitro single-unit
Ihab H Hassan et al.
The Journal of biological chemistry, 287(7), 4679-4689 (2011-12-24)
Known therapies for influenza A virus infection are complicated by the frequent emergence of resistance. A therapeutic strategy that may escape viral resistance is targeting host cellular mechanisms involved in viral replication and pathogenesis. The endoplasmic reticulum (ER) stress response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service