Skip to Content
MilliporeSigma

Skip To

580221

Taurodeoxycholic Acid, Sodium Salt

Detergent useful for the solubilization of lipids and membrane-bound proteins. Aggregation number: 6.

Synonym(s):

Taurodeoxycholic Acid, Sodium Salt, Sodium Taurodeoxycholate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H44NO6S · Na
CAS Number:
Molecular Weight:
521.69
MDL number:
UNSPSC Code:
41141802
NACRES:
NB.34
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (TLC)

form

solid

mol wt

521.28  g/mol

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

aggregation number

6

color

white

pKa 

1.9

CMC

1 - 4 mM

mp

171 -175 °C

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCNC(=O)CCC(C1C2(C(C3C(C4(C(CC3O)CC(CC4)O)C)CC2)CC1)C)C

InChI

1S/C26H45NO7S.Na/c1-16(4-7-23(30)27-12-13-34-35(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29;/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1

InChI key

XDQMFTUJGWFAGC-UHFFFAOYSA-M

General description

Detergent useful for the solubilization of lipids and membrane-bound proteins (aggregation number: 6; CMC: 1-4 mM; pKa: 1.9).

Application


  • Microemulsion electrokinetic chromatography of corticosteroids: Effect of surfactants and cyclodextrins on the separation selectivity.: This study illustrates the application of taurodeoxycholic acid, sodium salt in chromatographic techniques, specifically enhancing the selectivity and efficiency of corticosteroid analyses, which is crucial for pharmaceutical quality control and research (Pomponio R et al., 2005).

  • Modified micellar electrokinetic chromatography in the analysis of catechins and xanthines in chocolate.: Demonstrates the utility of taurodeoxycholic acid, sodium salt in improving the analysis of bioactive compounds in food products, facilitating better understanding of food composition and potential health benefits (Gotti R et al., 2004).

  • Separation and determination of effective components in silymarin and preparations of silymarin by MECC.: This research uses taurodeoxycholic acid, sodium salt to enhance the micellar electrokinetic chromatography of silymarin, a milk thistle extract known for its liver-protective effects, underscoring the role of advanced analytical techniques in herbal medicine (Ding TM et al., 2000).

Packaging

Packaged under inert gas

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T0557T0875900073
description

Detergent useful for the solubilization of lipids and membrane-bound proteins. Aggregation number: 6.

description

BioXtra, ≥97% (TLC)

description

≥95% (HPLC)

description

≥98 atom % D, ≥98% (CP)

assay

≥95% (TLC)

assay

≥97% (TLC)

assay

≥95% (HPLC)

assay

≥98% (CP)

form

solid

form

-

form

-

form

powder

aggregation number

6

aggregation number

6

aggregation number

6

aggregation number

-

storage temp.

10-30°C

storage temp.

-

storage temp.

-

storage temp.

−20°C

mol wt

521.28  g/mol

mol wt

micellar avg mol wt 3100

mol wt

micellar avg mol wt 3100

mol wt

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Taurodeoxycholic Acid, Sodium Salt


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters