Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559410

Sigma-Aldrich

SB 290157

A non-peptide that acts as a selective, high affinity, competitive antagonist of the anaphylatoxin C3a receptor (IC₅₀ = 200 nM).

Synonym(s):

SB 290157, N²-[(2,2-Diphenylethoxy)acetyl]-L-arginine, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N4O4 · xC2HF3O2
CAS Number:
Molecular Weight:
412.48 (free base basis)
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FC(F)(F)C(=O)O.N([C@@H](CCCN\C(=N/[H])\N)C(=O)O)C(=O)COCC(c2ccccc2)c1ccccc1

InChI

1S/C22H28N4O4.C2HF3O2/c23-22(24)25-13-7-12-19(21(28)29)26-20(27)15-30-14-18(16-8-3-1-4-9-16)17-10-5-2-6-11-17;3-2(4,5)1(6)7/h1-6,8-11,18-19H,7,12-15H2,(H,26,27)(H,28,29)(H4,23,24,25);(H,6,7)/t19-;/m0./s1

InChI key

ZJRMPPVJAQWGEG-FYZYNONXSA-N

General description

A non-peptide that acts as a selective, high affinity, competitive antagonist of the anaphylatoxin C3a receptor (IC50 = 200 nM). Does not antagonize C5aR or other chemotactic G-protein-coupled receptors. Blocks C3a-induced internalization of C3aR in human neutrophils and C3a-induced Ca2+ mobilization in basophilic leukemia RBL-2H3 cells expressing murine, guinea pig, or human C3aR (IC50 = 7, 12.5, and 27.7 nM, respectively). Also blocks C3a-mediated ATP release from Guinea pig platelets (IC50 = 30 nM). Exhibits anti-inflammatory properties. Supplied as a trifluoroacetate salt.
A non-peptide with anti-inflammatory properties that acts as a selective, high affinity, competitive antagonist of the anaphylatoxin C3a receptor (C3aR; IC50 = 200 nM). Does not antagonize C5aR or other chemotactic G-protein coupled receptors. Blocks C3a-induced internalization of C3aR in human neutrophils and C3a-induced Ca2+ mobilization in basophilic leukemia RBL-2H3 cells expressing murine, guinea pig, or human C3aR (IC50 = 7, 12.5, and 27.7 nM, respectively). Also blocks C3a-mediated ATP release from guinea pig platelets (IC50 = 30 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
C3a receptor
Product does not compete with ATP.
Reversible: no
Target IC50: 200 nM against the anaphylatoxin C3a receptor; 7, 12.5, and 27.7 nM, in blocking C3a-induced internalization of C3aR in human neutrophils and C3a-induced Ca2+ mobilization in basophilic leukemia RBL-2H3 cells expressing murine, guinea pig, or human C3aR

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Do not reuse solutions diluted in physiological buffers.

Other Notes

Ames, R.S., et al. 2001. J. Immunol. 166, 6341.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service