Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33797

Sigma-Aldrich

C5aR Antagonist, DF2593A

Synonym(s):

C5aR Antagonist, DF2593A, (R)-4-(1-(4-(piperidin-1-yl)butanamido)ethyl)phenyl trifluoromethanesulfonate, C5aR Inhibitor, DF2593A, C5aR Blocker, DF2593A, C5a Receptor Antagonist, DF2593A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25F3N2O4S
CAS Number:
Molecular Weight:
422.46
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

An orally available, blood-brain barrier permeant piperidinylbutanamide compound that acts as a highly potent, selective, non-competitive and allosteric antagonist of C5a receptor (C5aR). Shown to effectively block C5a-induced human PMN migration (IC50 = 5.0 nM) and is also effective in rat and mouse orthologs (IC50 = 6.0 and 1 nM for rat and mouse, respectively). Exhibits good selectivity against a panel of GPCRs and ion channels screened and is shown to be completely inactive in a prostaglandin E2 production assay (~10 µM). Blocks C5a-induced and carrageenan-induced hyperalgesia in mice without any sedative or central opioid-like effects. Following oral administration, it is rapidly absorbed and displays favorable pharmacokinetic profile (Cmax = 100 nM; Tmax = 1.2 h at 1 mg/kg in mice).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
C5aR

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Moriconi, A., et al. 2014. PNAS.111, 16937.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service