Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

557514

Sigma-Aldrich

Ro-31-8425

A potent, cell-permeable, reversible, ATP-competitive, and selective protein kinase C inhibitor (IC₅₀ = 15 nM for rat brain PKC).

Synonym(s):

Ro-31-8425, 2-[8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-3-yl]-3-(1-methyl-1H-indol-3-yl)maleimide, HCl, Bisindolylmaleimide X, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N4O2 · xHCl
CAS Number:
Molecular Weight:
424.49 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red to orange-red

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21c3c(c(c2CC(CC1)CN)C4=C(C(=O)NC4=O)c5c6c([n](c5)C)cccc6)cccc3

InChI

1S/C26H24N4O2/c1-29-14-18(16-6-2-4-8-19(16)29)23-24(26(32)28-25(23)31)22-17-7-3-5-9-20(17)30-11-10-15(13-27)12-21(22)30/h2-9,14-15H,10-13,27H2,1H3,(H,28,31,32)

InChI key

QQKKPWJFHBFCTH-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, ATP-competitive, and selective protein kinase C inhibitor (IC50 = 15 nM for rat brain PKC). Exhibits some degree of isozyme specificity (IC50 = 8 nM for PKCα, 8 nM for PKCβI, 14 nM for PKCβII, 13 nM for PKCγ, and 39 nM for PKCε). Shows slight selectivity for the conventional PKC isozymes PKCα, PKCβ, and PKCγ over the Ca2+-independent PKC isozyme PKCε. Upregulates CD11a expression and enhances mesenchymal stromal cell adhesion under flow.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product competes with ATP.
Reversible: yes
Target IC50: 15 nM for rat brain PKC; 8 nM for PKCα, 8 nM for PKCβI, 14 nM for PKCβII, 13 nM for PKCγ, and 39 nM for PKCε

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Levy, O., et al. 2015. Cell Reports10, In press.
Merritt, J.E., et al. 1997. Cell Signal 9, 53.
Wilkinson, S.E., et al. 1993. Biochem. J. 294, 335.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susheel N Chaurasia et al.
eLife, 11 (2022-10-04)
Notch signaling dictates cell fate decisions in mammalian cells including megakaryocytes. Existence of functional Notch signaling in enucleate platelets remains elusive. Transcripts/peptides of Notch1 and Delta-like ligand (DLL)-4 were detected in platelets isolated from human blood by RT-qPCR, Western analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service