Skip to Content
MilliporeSigma
All Photos(1)

Documents

203293

Sigma-Aldrich

Bisindolylmaleimide I

InSolution, ≥95%, protein kinase C (PKC) inhibitor

Synonym(s):

InSolution Bisindolylmaleimide I, 2-[1-(3-Dimethylaminopropyl)-1H-indol-3-yl]-3-(1H-indol-3-yl)-maleimide, Gö 6850

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N4O2
CAS Number:
Molecular Weight:
412.48
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C25H24N4O2/c1-28(2)12-7-13-29-15-19(17-9-4-6-11-21(17)29)23-22(24(30)27-25(23)31)18-14-26-20-10-5-3-8-16(18)20/h3-6,8-11,14-15,26H,7,12-13H2,1-2H3,(H,27,30,31)

InChI key

QMGUOJYZJKLOLH-UHFFFAOYSA-N

General description

A highly selective cell-permeable protein kinase C (PKC) inhibitor (Ki = 10 nM) that is structurally similar to staurosporine. Acts as a competitive inhibitor for the ATP-binding site of PKC. Shows high selectivity for PKCα, βI, βII, γ, δ, and ε isozymes. May inhibit protein kinase A at a much higher concentration (Ki = 2 µM). Also inhibits telomerase A treated NPC-076 cells. A 1 mg/ml solution of Bisindolylmaleimide I (Cat. No. 203290) in anhydrous DMSO.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product competes with ATP.
Reversible: yes
Target Ki: 10 nM against PKC

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 1 mg/ml solution of Bisindolylmaleimide I (Cat. No. 203290) in anhydrous DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Ku, W.-C., et al. 1997. Biochem. Biophys. Res. Commun.241, 730.
Gekeler, V., et al. 1996. Br. J. Cancer74, 897.
Kiss, Z., et al. 1995. Biochim. Biophys. Acta1265, 93.
Muid, R.E., et al. 1991. FEBS Lett.293, 160.
Toullec, D., et al. 1991. J. Biol. Chem.266, 15771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service