Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

189490

Sigma-Aldrich

3-MA

≥95% (HPLC), powder, autophagy inhibitor, Calbiochem®

Synonym(s):

Autophagy Inhibitor, 3-MA, 3-Methyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N5
CAS Number:
Molecular Weight:
149.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Autophagy Inhibitor, 3-MA, Autophagy Inhibitor, 3-MA, CAS 5142-23-4, is a cell-permeable autophagic sequestration blocker. Acts as an inhibitor of III PI3-Kinase.

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to faint yellow

solubility

DMSO: 10 mg/mL
water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1(C2=NC=NC2=C(N=C1)N)C

InChI

1S/C6H7N5/c1-11-3-10-5(7)4-6(11)9-2-8-4/h2-3H,7H2,1H3

InChI key

FSASIHFSFGAIJM-UHFFFAOYSA-N

General description

A widely used cell-permeable autophagic sequestration blocker that effectly protects cerebellar granule cells from apoptosis following serum/potassium deprivation. Although a known class III PI3K inhibitor (IC50 = 4.5 mM in cell-free enzymatic assays), 3-MA exhibits quite distinct pharmacological properties from those of two other PI3K inhibitors, LY 294002 (Cat. Nos. 440202 and 440204) and Wortmannin (Cat. No. 681675)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

In applications where high working concentrations are necessary (e.g. 10 mM), the use of a pre-made stock solution may result in the introduction of higher than desired solvent (e.g., DMSO) into the experimental systems. In such cases, it is recommended that the solid compound be weighed out at the time of use and reconstituted directly into the solutions used in the experimental system (buffers, media, etc.). Vortexing may be required for complete solubilization in water.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Canu, N., et al. 2005. J. Neurochem.92, 1228.
Hirosako, K., et al. 2004. Biochem. Biophys. Res. Commun.316, 845.
Beugnet, A., et al. 2003. Biochem. J.372, 555.
Eskelinen, E.L., et al. 2002. Traffic3, 878.
Petiot, A., et al. 2000. J. Biol. Chem.275, 992.
Seglen, P.O. and Gordon, P.B., 1982. Proc. Natl. Acad. Sci. USA79, 1889.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong Chen et al.
Cell death & disease, 13(2), 150-150 (2022-02-16)
Ferroptosis, which is characterized by intracellular iron accumulation and lipid peroxidation, is a newly described form of regulated cell death that may play a key role in tumour suppression. In the present study, we investigated the expression profiles and biological
Jinghui Zhai et al.
Journal of biochemical and molecular toxicology, 35(11), e22896-e22896 (2021-08-24)
The NOD-like receptor family pyrin domain-containing (NLRP3) inflammasomes is centrally implicated in cisplatin (CP)-induced kidney injury. Autophagy is critical for inhibiting production of NLRP3 protein that effectively reduces the inflammatory response. Ginsenoside Rg3 (SY), an active component extracted from ginseng
Wei Huang et al.
Frontiers in neuroscience, 15, 745815-745815 (2021-12-07)
Parkinson's disease is a neurodegenerative disorder with an inflammatory response as the core pathogenic mechanism. Previous human genetics findings support the view that the loss of TREM2 function will aggravate neurodegeneration, and TREM2 is one of the most highly expressed
Halima Alsamri et al.
Frontiers in oncology, 12, 911615-911615 (2022-06-18)
We recently reported that carnosol induces ROS-dependent autophagy and apoptosis in breast cancer cells. We also reported that carnosol inhibits breast cancer cell migration, invasion, and in ovo tumor growth, as well as targets STAT3, PCAF, and p300 to proteasome
Abdelhabib Semlali et al.
Frontiers in oncology, 12, 873447-873447 (2022-10-04)
Treatment of oral cancer is based exclusively on surgery combined with or without chemotherapy. However, it has several side effects. Targeting a new, more effective therapy has become an urgent matter. The purpose of this study was to evaluate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service