Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538540

Sigma-Aldrich

MMP-7, Proenzyme, Human, Recombinant, E. coli

Synonym(s):

Promatrilysin, rhPro-MMP-7

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.77

form

liquid

Quality Level

specific activity

≥1400 units/mg protein (after activation)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human pro-MMP-7 expressed in E. coli. MMP-7 is believed to be a key factor in the regulation of metastasis. Overexpression of MMP-7 is observed in colon cancer. Activatable by organomercurials (APMA; Cat. No. 164610).

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will digest 1.0 µg AZOCOLL Substrate (Cat. No. 194933), in the presence of 500 μM p-aminophenylmercuric acetate (APMA; Cat. No. 164610) per minute at 37°C, pH 7.5.

Physical form

In 150 mM NaCl, 25 mM Tris-HCl, 5 mM CaCl₂, 0.02% NaN₃, 0.01% Brij® 35 Detergent, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Freeze-thaw cycles may accelerate the activation process.

Other Notes

Girolamo, N. D., et al. 2001. Invest. Ophtal. Visual. Sci.42, 1963.
Kihira, Y., et al. 1996. Urol. Oncol. 2, 20.
Woessner, J.F. 1995. Methods Enzymol. 248, 485.
Crabbe, T., et al. 1992. Biochemistry 31, 8500.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service