Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444270

Sigma-Aldrich

MMP-7, Active, Human, Recombinant, E. coli

Synonym(s):

Matrilysin, rhMMP-7, PUMP-1

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.77

form

liquid

Quality Level

specific activity

≥3000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human MMP-7 expressed in E. coli. MMP-7, the smallest member of the matrix metalloproteinase family that can degrade a wide range of gelatins, proteoglycans, and glycoproteins of the matrix and can activate several other MMPs.
Recombinant, human MMP-7 expressed in E. coli. MMP-7 is the smallest member of the matrix metalloproteinase family that can degrade a wide range of gelatins, proteoglycans, and glycoproteins of the matrix and activate several other MMPs.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will digest 1.0 µg AZOCOLL Substrate (Cat. No. 194933) per min at 37°C, pH 7.5.

Physical form

In 150 mM NaCl, 10 mM HEPES, 5 mM CaCl₂, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kivelä-Rajamäki, M., et al. 2003. J. Periodont. Res.38, 583.
Woessner, J.F. 1995. Methods Enzymol. 248, 485.
Crabbe, T., et al. 1992. Biochemistry 31, 8500.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott A Johnson et al.
Methods in molecular medicine, 105, 95-106 (2004-10-20)
The primary sites of hematopoiesis change during murine ontogeny. The first blood cells emerge in two waves in the yolk sac; primitive erythroblasts, megakaryocytes, and macrophages emerge on embryonic d (E) 7.0, whereas definitive progenitor cells appear as clusters within
Katherine Conant et al.
Journal of neuroinflammation, 14(1), 56-56 (2017-03-18)
Matrix metalloproteinases (MMPs) are a family of enzymes that are typically released from intracellular stores to act on specific extracellular substrates. MMP expression and activity can be increased in a neuronal activity-dependent manner, and further increased in response to tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service