Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.38285

Sigma-Aldrich

Glucosylceramide Synthase Inhibitor, Genz-123346

Synonym(s):

Glucosylceramide Synthase Inhibitor, Genz-123346, (1R,2R)-nonanoic acid[2-(2ʹ,3ʹ-dihydro-benzo [1, 4] dioxin-6ʹ-yl)-2-hydroxy-1-pyrrolidin-1-ylmethyl-ethyl]-amide, GL1 synthase Inhibitor, Genz-123346, Genz123346

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38N2O4
CAS Number:
Molecular Weight:
418.57
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

InChI

1S/C24H38N2O4/c1-2-3-4-5-6-7-10-23(27)25-20(18-26-13-8-9-14-26)24(28)19-11-12-21-22(17-19)30-16-15-29-21/h11-12,17,20,24,28H,2-10,13-16,18H2,1H3,(H,25,27)/t20-,24-/m1/s1

InChI key

JMNXWOFCUJJYEO-HYBUGGRVSA-N

General description

A cell-permeable, orally bioavailable fatty acid amide derivative that acts as a highly potent and specific inhibitor of Glucosylceramide Synthase (GL1 synthase; IC50 = 14 nM for ganglioside GM1 inhibition). Does not affect the activities of other related glycosidase, such as 1-O-acylceramide synthase, a-glucosidase, and glucocerebrosidase in any significant manner. Shown to improve glucose tolerance, improve insulin sensitivity, and normalize hemoglobin A1C levels in Zucker diabetic fatty rats (75 mg/kg/day, p.o. for 6 weeks). Also acts as an inhibitor of GlcCer synthase and blocks the conversion of ceramide to GlcCer in murine models that are orthologous to human autosomal dominant polycystic kidney disease and nephronophthisis. Diminishes Akt-mTOR signaling by reducing phosphorylation of Akt and ribosomal S6 kinase in HEK293T cells without affecting their total levels.
A cell-permeable, orally bioavailable fatty acid amide derivative that acts as a highly potent and specific inhibitor of Glucosylceramide Synthase (GL1 synthase; IC50 = 14 nM for ganglioside GM1 inhibition). Does not affect the activities of other related glycosidase, such as 1-O-acylceramide synthase, a-glucosidase, and glucocerebrosidase in any significant manner. Shown to improve glucose tolerance, improve insulin sensitivity, and normalize hemoglobin A1C levels in Zucker diabetic fatty rats (75 mg/kg/day, p.o. for 6 weeks). Also acts as an inhibitor of GlcCer synthase and blocks the conversion of ceramide to GlcCer in murine models that are orthologous to human autosomal dominant polycystic kidney disease and nephronophthisis. Diminishes Akt-mTOR signaling by reducing phosphorylation of Akt and ribosomal S6 kinase in HEK293T cells without affecting their total levels.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
glucosylceramide synthase
Reversible: yes
Target IC50: 14 nM for ganglioside GM1 suppression

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shen, W., et al. 2014. J. Neurochem.129, 884.

Natoli, T.A., et al. 2010. Nat. Med.16, 788.

Zhao, H., et al. 2007. Diabetes.56, 1210.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service