Skip to Content
MilliporeSigma

SML0612

Perifosine

≥98% (HPLC), Protein kinase B/Akt inhibitor, powder

Synonym(s):

4-[[Hydroxy(octadecyloxy)phosphinyl]oxy]-1,1-dimethyl-piperidinium inner salt, KRX-0401, Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H52NO4P
CAS Number:
Molecular Weight:
461.66
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Perifosine, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC1CC[N+](CC1)(C)C)OCCCCCCCCCCCCCCCCCC

InChI

1S/C25H52NO4P/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-24-29-31(27,28)30-25-20-22-26(2,3)23-21-25/h25H,4-24H2,1-3H3

InChI key

SZFPYBIJACMNJV-UHFFFAOYSA-N

Application

Perifosine has been used as an alkyl phospholipid AKT inhibitor.

Biochem/physiol Actions

Perifosine (KRX-0401) is a selective bioavailable alkylphospholipid inhibitor of protein kinase B/Akt (PKB/Akt) with anti-proliferative activity.
Perifosine (KRX-0401) is a selective bioavailable alkylphospholipid inhibitor of protein kinase B/Akt (PKB/Akt) with anti-proliferative activity. Perifosine acts on cell membranes, selectively targeting proliferating cells, inducing growth arrest and apoptosis.
Perifosine (octadecyl-(1,1-dimethyl-4-piperidylio)) is an antitumor compound. It acts at lipid rafts and stops lysosomal accumulation and mTORC1 (mammalian target of rapamycin complex 1) signaling. This drug exhibits significant antiproliferative activity in vitro and in vivo in various human cancer model systems.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library