Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30339

Sigma-Aldrich

(+)-Abscisic Acid

≥98% (HPLC), powder, growth regulator, Calbiochem

Synonym(s):

(+)-Abscisic Acid, (+)-cis,trans-Abscisic Acid, S-ABA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Product Name

(+)-Abscisic Acid,

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

SMILES string

O[C@]1(C(CC(=O)C=C1C)(C)C)\C=C\C(=C/C(=O)O)\C

InChI

1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-/t15-/m1/s1

InChI key

JLIDBLDQVAYHNE-YKALOCIXSA-N

General description

An isoprenoid plant hormone that acts as a growth regulator affecting leaf and fruit abscission and increases permeability of plant cells to water. Under osmotic stress conditions it alters the osmotic potential of stomatal guard cells, causing them to shrink and stomata to close. Also acts as a proton pump inhibitor. The trans form is shown to potentiate NMDA-gated currents in hippocampal neurons (EC50 = 440 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PYR/PYL/RCAR

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jacob, T., et al. 1999. PNAS96, 12192.
Cutler, S.R., et al. 2010. Annu Rev Plant Biol.61, 651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service